Fumaroyl chloride 95%

Supplier: Thermo Scientific
Danger

Synonyms: Fumaryl chloride

A14540.14 A14540.22
A14540.14EA 61.1 EUR
A14540.14 A14540.22
Fumaroyl chloride 95%
Fumaroyl chloride

Purity: 95% CAS No.: 627-63-4 Molecular Formula: C4H2Cl2O2 Formula Weight: 152.97 Flash Point: 73°(163°F) Refractive Index: 1.5000 Odour: Acrid Melting Point: -2° Boiling Point: 159-161° Beilstein Reference: 1721342 UN No.: 1780 MDL No.: MFCD00000733

Formula: C₄H₂Cl₂O₂
MW: 152,96 g/mol
Boiling Pt: 159…161 °C
Melting Pt: –2 °C
Density: 1,413
Flash Pt: 73 °C (163 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000733
CAS Number: 627-63-4
EINECS: 211-005-4
UN: 1780
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR