Dimethyl fumarate ≥99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Fumaric acid dimethyl ester

A10402.14 A10402.22 A10402.30
A10402.14EA 33.7 EUR
A10402.14 A10402.22 A10402.30
Dimethyl fumarate ≥99%
Dimethyl fumarate

Purity: 99% CAS No.: 624-49-7 Molecular Formula: C6H8O4 Formula Weight: 144.13 Melting Point: 102-105° Boiling Point: 192-193° Merck Reference: 14,4287 MDL No.: MFCD00064438

Formula: C₆H₈O₄
Boiling Pt: 192…193 °C
Melting Pt: 102…105 °C
Density: 1,37
Storage Temperature: Ambient
MDL Number: MFCD00064438
CAS Number: 624-49-7
EINECS: 210-849-0
Merck Index: 14,4287

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR