Maleic acid
Supplier: Molekula
|
Warning
|
MOLE37286853-5KGEA
291
EUR
MOLE37286853-5KG
Maleic acid
Maleic acid
Formula:
C₄H₄O₄ MW: 116,07 g/mol Boiling Pt: 256 °C (1013 hPa) Melting Pt: 130…131 °C Density: 1,59 g/cm³ (20 °C) Flash Pt: 127 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00063177 CAS Number: 110-16-7 EINECS: 203-742-5 UN: 3261 ADR: 8,III Merck Index: 13,05726 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...