3-Methyl-2-buten-1-ol 99%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 2-Methyl-2-buten-4-ol

171920050 171921000 171925000
ACRO171920050EA 31.7 EUR
ACRO171920050 ACRO171921000 ACRO171925000
3-Methyl-2-buten-1-ol 99%
3-Methyl-2-buten-1-ol
Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 140
Melting Pt: –86 °C
Density: 0.84 g/cm³
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00002916
CAS Number: 556-82-1
EINECS: 209-141-4
UN: 1987
ADR: 3 III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4410 to 1.4450 (20°C, 589 nm)
GC ≥98.5 %
Specific gravity (25°C) 0.857 to 0.867
Specific optical rotation -8.5° to -10.5° (20°C, 589 nm) (c=1, water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR