3,4-Dichlorocinnamic acid ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 3,4-Dichlorocinnamic Acid

B25308.06 B25308.14
B25308.06EA 36.8 EUR
B25308.06 B25308.14
3,4-Dichlorocinnamic acid ≥97%
3,4-Dichlorocinnamic acid

Purity: 97% CAS No.: 1202-39-7 Molecular Formula: C9H6Cl2O2 Formula Weight: 217.05 Melting Point: 218-220° Beilstein Reference: 1872129

Formula: C₉H₆Cl₂O₂
MW: 217,05 g/mol
Melting Pt: 218…220 °C
Storage Temperature: Ambient
MDL Number: MFCD00004385
CAS Number: 1202-39-7
EINECS: 214-866-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR