Sinapic acid

Supplier: Roth Carl
Warning

Synonyms: 3,5-Dimethoxy-4-hydroxycinnamic acid, 3',5'-Dimethoxy-4'-hydroxycinnamic acid, 4-Hydroxy-3,5-dimethoxycinnamic acid

5317.1 5317.2
ROTH5317.1EA 105 EUR
ROTH5317.1 ROTH5317.2
Sinapic acid
Sinapic acid
Formula: C₁₁H₁₂O₅
MW: 224,21 g/mol
Boiling Pt: 378 °C (1013 hPa)
Melting Pt: 200…202 °C
MDL Number: MFCD00004401
CAS Number: 530-59-6
EINECS: 208-487-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR