trans-m-Coumaric acid ≥99%

Supplier: Thermo Scientific
Warning

Synonyms: (2E)-3-(3-Hydroxyphenyl)acrylic acid, m-Hydroxycinnamic acid, predominantly trans, trans-3-Hydroxycinnamic acid, (E)-m-Coumaric acid, (E)-3-(3-Hydroxyphenyl)acrylic acid

A14466.09 A14466.18 A14466.22
A14466.09EA 30.8 EUR
A14466.09 A14466.18 A14466.22
trans-m-Coumaric acid ≥99%
trans-m-Coumaric acid

Purity: 99% CAS No.: 14755-02-3 Molecular Formula: C9H8O3 Formula Weight: 164.16 Melting Point: 192-196° Beilstein Reference: 2690254

Formula: C₉H₈O₃
MW: 164,16 g/mol
Melting Pt: 192…196 °C
Storage Temperature: Ambient
MDL Number: MFCD00004390
CAS Number: 14755-02-3
EINECS: 209-615-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR