1-Bromo-2-butyne ≥98%
Supplier: Thermo Scientific
|
Warning
|
L20134.06
L20134.14
L20134.06EA
178
EUR
L20134.06
L20134.14
1-Bromo-2-butyne ≥98%
1-Bromo-2-butyne
Purity: 98%
CAS No.: 3355-28-0
Molecular Formula: CH3C≡CCH2Br
Formula Weight: 132.99
Flash Point: 36°(96°F)
Refractive Index: 1.5080
Boiling Point: 123-125°
Beilstein Reference: 605306
UN No.: 1993
MDL No.: MFCD00190233
Formula:
C₄H₅Br MW: 132,99 g/mol Boiling Pt: 123…125 °C Density: 1,519 Flash Pt: 36 °C (96 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00190233 CAS Number: 3355-28-0 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...