5-Hexynoic acid 97%
Supplier: Thermo Scientific
|
Danger
|
383700010
383700050
383700250
ACRO383700010EA
49.5
EUR
ACRO383700010
ACRO383700050
ACRO383700250
5-Hexynoic acid 97%
5-Hexynoic acid
Formula:
C₆H₈O₂ MW: 112,13 g/mol Boiling Pt: 221 °C (1013 hPa) Melting Pt: 41 °C Density: 1,016 g/cm³ Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00066346 CAS Number: 53293-00-8 UN: 2922 ADR: 8,II |
Specification Test Results
Appearance | Clear yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4480 to 1.4500 (20°C, 589 nm) |
Titration with NaOH | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...