2-Heptyn-1-ol ≥97%

Supplier: Thermo Scientific
Warning

A14959.03 A14959.06 A14959.14
A14959.03EA 25.5 EUR
A14959.03 A14959.06 A14959.14
2-Heptyn-1-ol ≥97%
2-Heptyn-1-ol

Purity: 97% CAS No.: 1002-36-4 Molecular Formula: CH3(CH2)3C≡CCH2OH Formula Weight: 112.17 Flash Point: 78°(172°F) Refractive Index: 1.4540 Boiling Point: 89-90°/16mm Beilstein Reference: 1739001 MDL No.: MFCD00039541

Formula: C₇H₁₂O
MW: 112,17 g/mol
Boiling Pt: 89…90 °C (16 mmHg)
Density: 0,882
Flash Pt: 78 °C (172 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039541
CAS Number: 1002-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR