3-Heptyn-1-ol ≥98%

Supplier: Thermo Scientific
Warning

B20884.06 B20884.14
B20884.06EA 48.5 EUR
B20884.06 B20884.14
3-Heptyn-1-ol ≥98%
3-Heptyn-1-ol

Purity: 98% CAS No.: 14916-79-1 Molecular Formula: CH3CH2CH2C≡CCH2CH2OH Formula Weight: 112.17 Flash Point: 74°(165°F) Refractive Index: 1.4555 Boiling Point: 80-82°/15mm Beilstein Reference: 1737178 MDL No.: MFCD00039551

Formula: C₇H₁₂O
MW: 112,17 g/mol
Boiling Pt: 80…82 °C (15 mmHg)
Density: 0,882
Flash Pt: 74 °C (165 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039551
CAS Number: 14916-79-1
EINECS: 238-985-6
UN: 1987
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR