N-Chlorosuccinimide 97%

Supplier: Thermo Scientific
Danger

Synonyms: 1-Chloropyrrolidine-2,5-dione

149251000 149255000 149250025
ACRO149251000EA 36.8 EUR
ACRO149251000 ACRO149255000 ACRO149250025
N-Chlorosuccinimide 97%
N-Chlorosuccinimide
Formula: C₄H₄ClNO₂
MW: 133,53 g/mol
Boiling Pt: 216,5 °C (1013 hPa)
Melting Pt: 144…150 °C
Density: 1,65 g/cm³ (20 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00005511
CAS Number: 128-09-6
EINECS: 204-878-8
UN: 3261
ADR: 8,II
Merck Index: 13,02183

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 144 to 150 °C
Titration Iodometric ≥96.0 % (on dry substance)
Loss on drying ≤4 % (2 g, 110 °C, 2 hrs)
Specific optical rotation +20.0° to +25.0° (20 °C, 589 nm) (c=1 in 8:2 AcOH/1M HCl)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR