Perhydroazepine 99%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Hexamethyleneimine
120590050
120595000
120590250
ACRO120590050EA
22.3
EUR
ACRO120590050
ACRO120595000
ACRO120590250
Perhydroazepine 99%
Perhydroazepine
Formula:
C₆H₁₃N MW: 99,18 g/mol Boiling Pt: 138 °C (760 mmHg) Melting Pt: –37 °C Density: 0,88 g/cm³ Flash Pt: 18 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006934 CAS Number: 111-49-9 EINECS: 203-875-9 UN: 2493 ADR: 3,II |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4650 to 1.4670 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...