Perhydroazepine ≥98%

Supplier: Thermo Scientific
Danger

Synonyms: Hexamethyleneimine

B24007.AE B24007.AP
B24007.AEEA 30.4 EUR
B24007.AE B24007.AP
Perhydroazepine ≥98%
Perhydroazepine

Purity: 98+% CAS No.: 111-49-9 Molecular Formula: C6H13N Formula Weight: 99.18 Flash Point: 22°(71°F) Refractive Index: 1.4660 Odour: Ammonia-like Melting Point: -37° Boiling Point: 138-139° Beilstein Reference: 1084 UN No.: 2493 MDL No.: MFCD00006934

Formula: C₆H₁₃N
MW: 99,18 g/mol
Boiling Pt: 138…139 °C
Melting Pt: –37 °C
Density: 0,880
Flash Pt: 22 °C (71 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006934
CAS Number: 111-49-9
EINECS: 203-875-9
UN: 2493
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR