4-Methylpiperidine 99%

Supplier: Thermo Fisher Scientific
Danger

127511000 127515000 127510025
ACRO127511000EA 40.8 EUR
ACRO127511000 ACRO127515000 ACRO127510025
4-Methylpiperidine 99%
4-Methylpiperidine
Formula: C₆H₁₃N
Boiling Pt: 122 °C (760 mmHg)
Melting Pt: 4…5 °C
Density: 0,84 g/cm³
Flash Pt: 13 °C
Storage Temperature: Ambient
MDL Number: MFCD00006005
CAS Number: 626-58-4
EINECS: 210-954-1
UN: 3286
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4440 to 1.4480 (20°C, 589 nm)
GC ≥98.5 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR