Azulene 99%

Supplier: Thermo Fisher Scientific

105180010 105181000
ACRO105180010EA 243 EUR
ACRO105180010 ACRO105181000
Azulene 99%
Azulene
Formula: C₁₀H₈
Boiling Pt: 242 °C (1013 hPa)
Melting Pt: 99…101 °C
MDL Number: MFCD00003810
CAS Number: 275-51-4
EINECS: 205-993-6
ADR: 9,III
Merck Index: 13,00928

Order Now

Specification Test Results

Appearance Dark blue Crystalline powder
Infrared spectrum Conforms
Melting point 99 to 101 °C
GC ≥98.5 %
Suitability (for use as a laser dye) Passes test
Molar absorption coefficient ≥20000 l/(mol.cm) (at Lambda max)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR