2-Indolinone ≥97%

Supplier: Thermo Fisher Scientific
Warning

A18764.22 A18764.06 A18764.14
A18764.22EA 409 EUR
A18764.22 A18764.06 A18764.14
2-Indolinone ≥97%
2-Indolinone

Purity: 97+% CAS No.: 59-48-3 Molecular Formula: C8H7NO Formula Weight: 133.15 Melting Point: 124-128° Boiling Point: 194-196°/17mm MDL No.: MFCD00005711

Formula: C₈H₇NO
Boiling Pt: 194…196 °C (17 mmHg)
Melting Pt: 124…128 °C
Density: 1,32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005711
CAS Number: 59-48-3
EINECS: 200-429-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR