5-Nitroisatin ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 5-Nitro-1H-indole-2,3-dione, 5-Nitroindoline-2,3-dione

B24962.06 B24962.14
B24962.06EA 40 EUR
B24962.06 B24962.14
5-Nitroisatin ≥98%
5-Nitroisatin

Purity: 98% CAS No.: 611-09-6 Molecular Formula: C8H4N2O4 Formula Weight: 192.13 Melting Point: ca 252° dec. Beilstein Reference: 180223

Formula: C₈H₄N₂O₄
MW: 192.13 g/mol
Melting Pt: ∼ 252 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005720
CAS Number: 611-09-6
EINECS: 210-252-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR