5,7-Dimethylisatin 96%

Supplier: Thermo Fisher Scientific

L11332.06 L11332.14
L11332.06EA 146 EUR
L11332.06 L11332.14
5,7-Dimethylisatin 96%
5,7-Dimethylisatin

Purity: 96% CAS No.: 39603-24-2 Molecular Formula: C10H9NO2 Formula Weight: 175.19 Melting Point: 244-248° Beilstein Reference: 143679

Formula: C₁₀H₉NO₂
Melting Pt: 244…248 °C
Storage Temperature: Ambient
MDL Number: MFCD00047219
CAS Number: 39603-24-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR