1,3,3-Trimethyl-2-methyleneindoline 95%

Supplier: Thermo Scientific
Warning

A16741.18 A16741.30
A16741.18EA 56.2 EUR
A16741.18 A16741.30
1,3,3-Trimethyl-2-methyleneindoline 95%
1,3,3-Trimethyl-2-methyleneindoline

Purity: 95% CAS No.: 118-12-7 Molecular Formula: C12H15N Formula Weight: 173.26 Flash Point: 101°(213°F) Refractive Index: 1.5780 Melting Point: -10° Boiling Point: 247-248° Beilstein Reference: 131162 MDL No.: MFCD00005813

Formula: C₁₂H₁₅N
MW: 173,26 g/mol
Boiling Pt: 247…248 °C
Melting Pt: –10 °C
Density: 0,979
Flash Pt: 101 °C (213 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005813
CAS Number: 118-12-7
EINECS: 204-235-1
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR