6-Nitropiperonal ≥98%
Supplier: Thermo Scientific
Synonyms:
6-Nitro-1,3-benzodioxole-5-carboxaldehyde
A14039.06
A14039.18
A14039.06EA
0
EUR
A14039.06
A14039.18
6-Nitropiperonal ≥98%
6-Nitropiperonal
Purity: 98+%
CAS No.: 712-97-0
Molecular Formula: C8H5NO5
Formula Weight: 195.13
Melting Point: 93-97°
Beilstein Reference: 384010
Formula:
C₈H₅NO₅ MW: 195,13 g/mol Melting Pt: 98 °C |
MDL Number:
MFCD00005829 CAS Number: 712-97-0 EINECS: 211-926-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...