Piperonyl alcohol 98%
Supplier: Thermo Fisher Scientific
Synonyms:
Benzo[d][1,3]dioxol-5-ylmethanol, 3,4-(Methylenedioxy)benzyl alcohol, 1,3-Benzodioxol-5-ylmethanol
131351000
131350250
ACRO131351000EA
138
EUR
ACRO131351000
ACRO131350250
Piperonyl alcohol 98%
Piperonyl alcohol
Formula:
C₈H₈O₃ MW: 152.15 g/mol Boiling Pt: 156…158 °C (66 mmHg) Melting Pt: 51…55 °C Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005836 CAS Number: 495-76-1 EINECS: 207-808-4 |
Specification Test Results
Appearance | White to yellow Crystalline powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 51 to 55 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...