4-Methoxyphenylboronic acid ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
4-Methoxybenzeneboronic acid, Dihydroxy(4-methoxyphenyl)borane, (4-Methoxyphenyl)metaboric acid
A14462.06
A14462.03
A14462.14
A14462.06EA
194
EUR
A14462.06
A14462.03
A14462.14
4-Methoxyphenylboronic acid ≥97%
4-Methoxyphenylboronic acid
Purity: 97+%
CAS No.: 5720-07-0
Molecular Formula: C7H9BO3
Formula Weight: 151.96
Melting Point: 202-208°
Beilstein Reference: 2936912
Formula:
C₇H₉BO₃ MW: 151,96 g/mol Melting Pt: 202…208 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039139 CAS Number: 5720-07-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...