3-Hydroxyphenylboronic acid 97%

Supplier: Thermo Scientific
Warning

Synonyms: (3-Hydroxyphenyl)metaboric acid, (3-Hydroxyphenyl)boronic acid, Dihydroxy(3-hydroxyphenyl)borane, 3-Hydroxybenzeneboronic acid

AC35941.250 359412500 359410010 359410100
MAYBAC35941.250EA 0 EUR
MAYBAC35941.250 ACRO359412500 ACRO359410010 ACRO359410100
3-Hydroxyphenylboronic acid 97%
3-Hydroxyphenylboronic acid
Formula: C₆H₇BO₃
MW: 137,93 g/mol
Melting Pt: 212 °C
Storage Temperature: Ambient
MDL Number: MFCD01074603
CAS Number: 87199-18-6

Order Now

Specification Test Results

Appearance Pink to grey to tan Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR