1-Bromo-2-chloro-5-fluorobenzene ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 2-Bromo-1-chloro-4-fluorobenzene

B25532.06 B25532.14
B25532.06EA 0 EUR
B25532.06 B25532.14
1-Bromo-2-chloro-5-fluorobenzene ≥98%
1-Bromo-2-chloro-5-fluorobenzene

Purity: 98+% CAS No.: 201849-15-2 Molecular Formula: C6H3BrClF Formula Weight: 209.45 Refractive Index: 1.5530

Formula: C₆H₃BrClF
MW: 209,44 g/mol
Density: 1,75
Storage Temperature: Ambient
MDL Number: MFCD00061120
CAS Number: 201849-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR