2-Bromobenzamide ≥98%

Supplier: Thermo Fisher Scientific
Warning

A18331.06 A18331.14
A18331.06EA 65.9 EUR
A18331.06 A18331.14
2-Bromobenzamide ≥98%
2-Bromobenzamide

Purity: 98% CAS No.: 4001-73-4 Molecular Formula: C7H6BrNO Formula Weight: 200.03 Melting Point: 160-162° MDL No.: MFCD00007969

Formula: C₇H₆BrNO
Boiling Pt: 198 °C (1013 hPa)
Melting Pt: 160…162 °C
Density: 1,609 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007969
CAS Number: 4001-73-4
EINECS: 223-650-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR