4-Bromobenzamide ≥97%

Supplier: Thermo Fisher Scientific
Warning

B24348.04 B24348.09 B24348.18
B24348.04EA 34.7 EUR
B24348.04 B24348.09 B24348.18
4-Bromobenzamide ≥97%
4-Bromobenzamide

Purity: 97% CAS No.: 698-67-9 Molecular Formula: C7H6BrNO Formula Weight: 200.03 Melting Point: 190-193° MDL No.: MFCD00007991

Formula: C₇H₆BrNO
Boiling Pt: 328 °C (1013 hPa)
Melting Pt: 190…193 °C
Storage Temperature: Ambient
MDL Number: MFCD00007991
CAS Number: 698-67-9
EINECS: 211-817-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR