1,4-Dibromotetrafluorobenzene ≥99%
Supplier: Thermo Fisher Scientific
|
Warning
|
A18135.03
A18135.06
A18135.14
A18135.03EA
43.3
EUR
A18135.03
A18135.06
A18135.14
1,4-Dibromotetrafluorobenzene ≥99%
1,4-Dibromotetrafluorobenzene
Purity: 99%
CAS No.: 344-03-6
Molecular Formula: C6Br2F4
Formula Weight: 307.87
Melting Point: 78-81°
MDL No.: MFCD00000309
Formula:
C₆Br₂F₄ MW: 307.87 g/mol Boiling Pt: 198 Melting Pt: 78…81 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000309 CAS Number: 344-03-6 EINECS: 206-448-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...