4-Bromo-2,3,5,6-tetrafluorobenzoyl chloride ≥98%

Supplier: Thermo Scientific
Danger

Synonyms: 4-Bromo-2,3,5,6-tetrafluorobenzene-1-carbonyl chloride

L20153.03 L20153.06
L20153.03EA 0 EUR
L20153.03 L20153.06
4-Bromo-2,3,5,6-tetrafluorobenzoyl chloride ≥98%
4-Bromo-2,3,5,6-tetrafluorobenzoyl chloride

Purity: 98% CAS No.: 122033-54-9 Molecular Formula: C7BrClF4O Formula Weight: 291.43 Refractive Index: 1.5170 Boiling Point: 95-100° UN No.: 3265 MDL No.: MFCD03001173

Formula: C₇BrClF₄O
MW: 291,43 g/mol
Boiling Pt: 95…100 °C
Storage Temperature: Ambient
MDL Number: MFCD03001173
CAS Number: 122033-54-9
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR