3-Bromothioanisole ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 3-Bromophenyl methyl sulphide, 1-Bromo-3-(methylthio)benzene

B25406.03 B25406.06
B25406.03EA 0 EUR
B25406.03 B25406.06
3-Bromothioanisole ≥98%
3-Bromothioanisole

Purity: 98% CAS No.: 33733-73-2 Molecular Formula: C7H7BrS Formula Weight: 203.11 Flash Point: 110°(230°F) Refractive Index: 1.6240 Boiling Point: 124-125°/10mm Beilstein Reference: 2078598 MDL No.: MFCD00041395

Formula: C₇H₇BrS
MW: 203,1 g/mol
Boiling Pt: 124…125 °C (10 mmHg)
Density: 1,510
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041395
CAS Number: 33733-73-2
UN: 3334
ADR: 9,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR