3,4-Dichlorophenylboronic acid 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
AC35909.1
AC35909.10
359090050
359090250
MAYBAC35909.1EA
0
EUR
MAYBAC35909.1
MAYBAC35909.10
ACRO359090050
ACRO359090250
3,4-Dichlorophenylboronic acid 97%
3,4-Dichlorophenylboronic acid
Formula:
C₆H₅BCl₂O₂ Melting Pt: 284…286 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01074646 CAS Number: 151169-75-4 |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Titration with NaOH | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...