2-Chloro-5-nitrobenzoyl chloride 96%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
2-Chloro-5-nitrobenzene-1-carbonyl chloride, 2-Chloro-5-nitrobenzoylchloride
L17523.14
L17523.22
L17523.14EA
64
EUR
L17523.14
L17523.22
2-Chloro-5-nitrobenzoyl chloride 96%
2-Chloro-5-nitrobenzoyl chloride
Purity: 96%
CAS No.: 25784-91-2
Molecular Formula: C7H3Cl2NO3
Formula Weight: 220.01
Melting Point: 58-60°
Boiling Point: 157-158°/11mm
UN No.: 3261
MDL No.: MFCD00059180
Formula:
C₇H₃Cl₂NO₃ MW: 220,01 g/mol Boiling Pt: 157…158 °C (11 mmHg) Melting Pt: 58…60 °C Storage Temperature: Ambient |
MDL Number:
MFCD00059180 CAS Number: 25784-91-2 EINECS: 247-262-4 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...