Benzoyl chloride 99%, pure
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Benzoic acid chloride
105750010
105750025
ACRO105750010EA
68
EUR
ACRO105750010
ACRO105750025
Benzoyl chloride 99%, pure
Benzoyl chloride
Formula:
C₇H₅ClO MW: 140,57 g/mol Boiling Pt: 197,2 °C (1013 hPa) Melting Pt: –1 °C Density: 1,21 g/cm³ Flash Pt: 93 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000653 CAS Number: 98-88-4 EINECS: 202-710-8 UN: 1736 ADR: 8,II Merck Index: 13,01113 |
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5520 to 1.5540 (20°C, 589 nm) |
GC | ≥98.5 % |
Color scale | ≤40 APHA |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...