2-Ethoxybenzoyl chloride ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
L17686.14
L17686.22
L17686.14EA
0
EUR
L17686.14
L17686.22
2-Ethoxybenzoyl chloride ≥98%
2-Ethoxybenzoyl chloride
Purity: 98%
CAS No.: 42926-52-3
Molecular Formula: C9H9ClO2
Formula Weight: 184.62
Refractive Index: 1.5530
Boiling Point: 114-118°/5mm
UN No.: 3265
MDL No.: MFCD00016348
Formula:
C₉H₉ClO₂ MW: 184.62 g/mol Boiling Pt: 114…118 °C (5 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD00016348 CAS Number: 42926-52-3 EINECS: 256-003-4 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...