2-Anisoyl chloride 97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 2-Methoxybenzoyl chloride, 2-Methoxybenzene-1-carbonyl chloride

276110050 276110250
ACRO276110050EA 56.7 EUR
ACRO276110050 ACRO276110250
2-Anisoyl chloride 97%
2-Anisoyl chloride
Formula: C₈H₇ClO₂
Boiling Pt: 128…129 °C (8 mmHg)
Density: 1,146 g/cm³
Flash Pt: 84 °C
MDL Number: MFCD00000664
CAS Number: 21615-34-9
EINECS: 244-477-5
UN: 1729
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5714 to 1.5734 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR