4-Anisoyl chloride ≥97%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
4-Methoxybenzoyl chloride
L13120.14
L13120.22
L13120.14EA
39.3
EUR
L13120.14
L13120.22
4-Anisoyl chloride ≥97%
4-Anisoyl chloride
Purity: 97%
CAS No.: 100-07-2
Molecular Formula: C8H7ClO2
Formula Weight: 170.60
Flash Point: 87°(188°F)
Refractive Index: 1.5810
Melting Point: 22°
Boiling Point: 145°/14mm
Beilstein Reference: 471918
Merck Reference: 14,673
UN No.: 1729
MDL No.: MFCD00000687
Formula:
C₈H₇ClO₂ Boiling Pt: 145 °C (14 mmHg) Melting Pt: 22 °C Density: 1,260 Flash Pt: 87 °C (188 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000687 CAS Number: 100-07-2 EINECS: 202-816-4 UN: 1729 ADR: 8,II Merck Index: 14,673 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...