Terephthaloyl dichloride ≥99%

Supplier: Thermo Scientific
Danger

Synonyms: Terephthaloyl chloride

A11224.0B A11224.30
A11224.0BEA 125 EUR
A11224.0B A11224.30
Terephthaloyl dichloride ≥99%
Terephthaloyl dichloride

Purity: 99% CAS No.: 100-20-9 Molecular Formula: C8H4Cl2O2 Formula Weight: 203.02 Flash Point: 180°(356°F) Melting Point: 79-83° Boiling Point: 265° Beilstein Reference: 607796 UN No.: 2928 MDL No.: MFCD00000693

Formula: C₈H₄Cl₂O₂
MW: 203,02 g/mol
Boiling Pt: 265 °C
Melting Pt: 79…83 °C
Density: 1,34 g/cm³ (100 °C)
Flash Pt: 180 °C (356 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000693
CAS Number: 100-20-9
EINECS: 202-829-5
UN: 2928
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR