Salicylamide 99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-Hydroxybenzamide

229360010 229360050 229362500
ACRO229360010EA 101 EUR
ACRO229360010 ACRO229360050 ACRO229362500
Salicylamide 99%
Salicylamide
Formula: C₇H₇NO₂
MW: 137.14 g/mol
Boiling Pt: 318
Melting Pt: 139…142 °C
MDL Number: MFCD00007978
CAS Number: 65-45-2
EINECS: 200-609-3
Merck Index: 12,08480

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 139 to 142 °C
Assay ≥98.5 % (on anhydrous substance)
Water ≤0.5 %
Sulfated ash ≤0.1 %
Heavy metals (as Pb) ≤10 ppm

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR