3-Nitrobenzhydrazide ≥98%

Supplier: Thermo Fisher Scientific
Warning

B20800.06 B20800.14
B20800.06EA 33.7 EUR
B20800.06 B20800.14
3-Nitrobenzhydrazide ≥98%
3-Nitrobenzhydrazide

Purity: 98+% CAS No.: 618-94-0 Molecular Formula: C7H7N3O3 Formula Weight: 181.15 Melting Point: 152-155° Beilstein Reference: 645512

Formula: C₇H₇N₃O₃
Melting Pt: 152…155 °C
Storage Temperature: Ambient
MDL Number: MFCD00017059
CAS Number: 618-94-0
EINECS: 210-572-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR