3,4,5-Trimethoxybenzhydrazide ≥98%
Supplier: Thermo Fisher Scientific
B20055.06
B20055.14
B20055.06EA
0
EUR
B20055.06
B20055.14
3,4,5-Trimethoxybenzhydrazide ≥98%
3,4,5-Trimethoxybenzhydrazide
Purity: 98%
CAS No.: 3291-03-0
Molecular Formula: C10H14N2O4
Formula Weight: 226.23
Melting Point: 158-160°
Beilstein Reference: 484531
Formula:
C₁₀H₁₄N₂O₄ Melting Pt: 158…160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014758 CAS Number: 3291-03-0 EINECS: 221-954-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...