Salicylhydrazide 98
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
2-Hydroxybenzoic acid hydrazide, Salicylohydrazide, 2-Hydroxybenzene-1-carbohydrazide, Salicylic acid hydrazide, 2-Hydroxybenzohydrazide, 2-Hydroxybenzhydrazide, 2-Hydroxybenzoic hydrazide
A14218.14
A14218.22
A14218.14EA
68.7
EUR
A14218.14
A14218.22
Salicylhydrazide 98
Salicylhydrazide
Purity: 98+%
CAS No.: 936-02-7
Molecular Formula: C7H8N2O2
Formula Weight: 152.15
Melting Point: 146-149°
Beilstein Reference: 472355
Formula:
C₇H₈N₂O₂ MW: 152.15 g/mol Melting Pt: 146…149 °C Density: 1.424 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007599 CAS Number: 936-02-7 EINECS: 213-311-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...