Diallyl phthalate 98%

Supplier: Thermo Scientific
Warning

Synonyms: Phthalic acid diallyl ester

276620010 276620025 276622500
ACRO276620010EA 78.7 EUR
ACRO276620010 ACRO276620025 ACRO276622500
Diallyl phthalate 98%
Diallyl phthalate
Formula: C₁₄H₁₄O₄
MW: 246,26 g/mol
Boiling Pt: 300 °C (760 mmHg)
Melting Pt: –70 °C
Density: 1,12 g/cm³
Flash Pt: 166 °C
Storage Temperature: Ambient
MDL Number: MFCD00008646
CAS Number: 131-17-9
EINECS: 205-016-3
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5180 to 1.5200 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR