Pentyl benzoate ≥98%

Supplier: Thermo Scientific

A10177.22 A10177.36
A10177.22EA 44.9 EUR
A10177.22 A10177.36
Pentyl benzoate ≥98%
Pentyl benzoate

Purity: 98+% CAS No.: 2049-96-9 Molecular Formula: C12H16O2 Formula Weight: 192.26 Flash Point: 130°(266°F) Refractive Index: 1.4950 Boiling Point: 260° MDL No.: MFCD00048843

Formula: C₁₂H₁₆O₂
MW: 192,26 g/mol
Boiling Pt: 260 °C
Density: 0,99
Flash Pt: 130 °C (266 °F)
Storage Temperature: Ambient
MDL Number: MFCD00048843
CAS Number: 2049-96-9
EINECS: 218-077-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR