Methyl salicylate 99%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
Wintergreen Oil, Methyl 2-hydroxybenzoate
220490010
220490050
220495000
220490025
ACRO220490010EA
121
EUR
ACRO220490010
ACRO220490050
ACRO220495000
ACRO220490025
Methyl salicylate 99%
Methyl salicylate
Formula:
2-(HO)C₆H₄CO₂CH₃ MW: 152.15 g/mol Boiling Pt: 222 °C (760 mmHg) Melting Pt: 8…7 °C Density: 1.1825 g/cm³ Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002214 CAS Number: 119-36-8 EINECS: 204-317-7 Merck Index: 13,06143 |
|
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5350 to 1.5380 (20°C, 589 nm) |
Density | (20/4°C) 1.180 to 1.186 g/ml |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...