Sodium benzoate

Supplier: Roth Carl

8548.1 8548.2
ROTH8548.1EA 33.5 EUR
ROTH8548.1 ROTH8548.2
Sodium benzoate
Sodium benzoate
Formula: C₆H₅COONa
Melting Pt: 410…430 °C
Density: 1,44 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00012463
CAS Number: 532-32-1
EINECS: 208-534-8
REACH: 01-2119460683-35
Merck Index: 13,08654

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR