4-Isopropoxybenzoic acid ≥99%
Supplier: Thermo Fisher Scientific
|
Warning
|
A14351.09
A14351.18
A14351.30
A14351.09EA
45.2
EUR
A14351.09
A14351.18
A14351.30
4-Isopropoxybenzoic acid ≥99%
Purity: 99%
CAS No.: 13205-46-4
Molecular Formula: C10H12O3
Formula Weight: 180.20
Odour: Odourless
Melting Point: 163-167°
MDL No.: MFCD00044318
Formula:
C₁₀H₁₂O₃ Boiling Pt: 305 °C (1013 hPa) Melting Pt: 163…167 °C Storage Temperature: Ambient |
MDL Number:
MFCD00044318 CAS Number: 13205-46-4 EINECS: 236-169-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...