Thiomersal

Supplier: Molekula
Danger

Synonyms: Ethylmercurithiosalicylic acid sodium salt, Sodium ethylmercurithiosalicylate, Thimerosal, Merthiolat, 2-[(Ethylmercury)thio]benzoic acid sodium salt, 2-(Ethylmercuriomercapto)benzoic acid sodium salt, 2-(Ethylmercuriothio)benzoic acid sodium salt

13841854-100G 13841854-25G
MOLE13841854-100GEA 755 EUR
MOLE13841854-100G MOLE13841854-25G
Thiomersal
Thiomersal
Formula: 2-(C₂H₅HgS)C₆H₄CO₂Na
Melting Pt: 232…233 °C
Flash Pt: 250 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00013062
CAS Number: 54-64-8
EINECS: 200-210-4
UN: 2025
ADR: 6.1,II
Merck Index: 12,09451
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=621fb9ecf6f0ee8129acf49c938a6167645828da&vid=a4d9481c20c7c69ffe3a642fbe6cae8c3dfb117f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR