Phthalic acid ≥99%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Pathalic acid
A14450.0B
A14450.0I
A14450.30
A14450.0BEA
109
EUR
A14450.0B
A14450.0I
A14450.30
Phthalic acid ≥99%
Phthalic acid
Purity: 99%
CAS No.: 88-99-3
Molecular Formula: C8H6O4
Formula Weight: 166.13
Flash Point: 168°(334°F)
Odour: Odourless
Beilstein Reference: 608199
Merck Reference: 14,7371
MDL No.: MFCD00002467
Formula:
C₈H₆O₄ MW: 166,13 g/mol Boiling Pt: 315…317 °C (1013 hPa) Melting Pt: 205 °C (dec.) Density: 1,593 Flash Pt: 168 °C (334 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002467 CAS Number: 88-99-3 EINECS: 201-873-2 Merck Index: 14,7371 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...