Isophthalic acid 99%
Supplier: Thermo Scientific
122650020
122650050
122655000
ACRO122650020EA
90
EUR
ACRO122650020
ACRO122650050
ACRO122655000
Isophthalic acid 99%
Isophthalic acid
Formula:
C₈H₆O₄ MW: 166,13 g/mol Boiling Pt: 119 - 121 °C (6 -rr) Melting Pt: 341…343 °C Density: 1,54 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002516 CAS Number: 121-91-5 EINECS: 204-506-4 Merck Index: 13,05216 |
Specification Test Results
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Sulfated ash | ≤0.0015 % |
Titration with NaOH | ≥98.5 % |
Cobalt (Co) | ≤2 ppm |
Iron (Fe) | ≤2 ppm |
Manganese (Mn) | ≤2 ppm |
Impurity | ≤0.0025 % 3-Carboxy benzaldehyde |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...