Tetrafluoroisophthalonitrile ≥97%

Supplier: Thermo Scientific
Danger

Synonyms: Tetrafluoro-1,3-benzenedicarbonitrile, Tetrafluorobenzene-1,3-dicarbonitrile, Tetrafluoro-1,3-dicyanobenzene

B21599.03 B21599.06 B21599.14
B21599.03EA 43.1 EUR
B21599.03 B21599.06 B21599.14
Tetrafluoroisophthalonitrile ≥97%
Tetrafluoroisophthalonitrile

Purity: 97% CAS No.: 2377-81-3 Molecular Formula: C8F4N2 Formula Weight: 200.10 Melting Point: 76-78° Beilstein Reference: 6925899 UN No.: 3439 MDL No.: MFCD00013290

Formula: C₈F₄N₂
MW: 200,1 g/mol
Boiling Pt: 283 °C (1013 hPa)
Melting Pt: 76…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00013290
CAS Number: 2377-81-3
EINECS: 000-000-0
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR